Learn More
Creative Biomart Recombinant Tick-Borne Encephalitis Virus gE Antigen
Supplier:  Creative Biomart TBE070
Encompass_Preferred
The E. coli derived recombinant 37 kDa protein NE contains the Tick-borne encephalitis virus N-terminus regions of glycoprotein E.
Catalog No. 50-208-4448
 Restricted Item