Learn More
Perkin Elmer US LLC Standard PVC Two-Stop Peristaltic Pump Tubing - 0.25 mm I.D., Orange/Blue, Pkg. 12
Catalog No. NC2039027
Encompass_Preferred
This non-flared PVC Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12. Flared PVC Pump Tubing is also available for Optima, Avio, and NexION.
Catalog No. NC2039027 Supplier Perkin Elmer US LLC Supplier No. N0773117
This non-flared PVC Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12. Flared PVC Pump Tubing is also available for Optima, Avio, and NexION.