Learn More
Accuflow Systems TBG BIOPHRM PLS 2 STP #16 8/PK
Catalog No. 501109096
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Tubing, Biopharm Plus, 2-stop, #16, 8/pk Specifications: Product Type - Stopped Pump Tubing Tubing formulation - BioPharm Plus (platinum cured silicone) Pump Series - L/S Masterflex tubing size (L/S) - 16
Catalog No. 50-110-9096 Supplier Accuflow Systems Supplier No. 9611616
Manufactured by Cole Parmer and sold by Accuflow. Tubing, Biopharm Plus, 2-stop, #16, 8/pk Specifications: Product Type - Stopped Pump Tubing Tubing formulation - BioPharm Plus (platinum cured silicone) Pump Series - L/S Masterflex tubing size (L/S) - 16