Learn More
Thermo Scientific Advanced nanoViper Fingertight Fitting Systems 20 μm | | 1 EA
Catalog No. NC2069632 Shop All Thermo Scientific Products
Encompass
Ensure optimal separations and maximize your liquid chromatography LC instrument performance with virtually zero-dead-volume connections. Advanced capillaries extend the use case based on application requirements.
Catalog No. NC2069632 Supplier Thermo Scientific Supplier No. 6041.5292
Ensure optimal separations and maximize your liquid chromatography LC instrument performance with virtually zero-dead-volume connections. Advanced capillaries extend the use case based on application requirements.