Learn More
Analytical West PVC 2-Stop Flared Tubing, Orange/Orange (0.89mm) 12/pk
SDP
Catalog No. NC1679853
Encompass
PVC Flow-measured tubing provides excellent pumping life, good handling characteristics and low permeability. Flared ends facilitate an easy connection between the sample and peristaltic pump tubing.
Catalog No. NC1679853 Supplier Analytical West Supplier No. PT2140PF
PVC Flow-measured tubing provides excellent pumping life, good handling characteristics and low permeability. Flared ends facilitate an easy connection between the sample and peristaltic pump tubing.