Learn More
Selleck Chemical LLC TBHQ-100mg
SDP
Catalog No. 502389844
Encompass_Preferred
Tert-butylhydroquinone (TBHQ) is an antioxidant compound which is used to prevent lipid peroxidation and shows multiple cytoprotective actions. It is an activator of Nrf2.
Catalog No. 50-238-9844 Supplier Selleck Chemical LLC Supplier No. S4990100MG
Tert-butylhydroquinone (TBHQ) is an antioxidant compound which is used to prevent lipid peroxidation and shows multiple cytoprotective actions. It is an activator of Nrf2.