Learn More
Medchemexpress LLC TBI-223 | 2071265-08-0 | 99.7% | 365.36 | 50 MG
SDP
Catalog No. 5000350329
Encompass_Preferred
TBI-223 is an orally active oxazolidinone antibiotic and an antimicrobial that demonstrates activity against Mycobacterium tuberculosis (Mtb). It exhibits an IC50 of 68 μg/mL for inhibiting mitochondrial protein synthesis (MPS) in HepG2 cells. It has shown effectiveness in three mouse models of methicillin-resistant Staphylococcus aureus infection, and can be utilized for the study of tuberculosis.
  • Orally active oxazolidinone antibiotic
  • Antimicrobial activity against Mycobacterium tuberculosis (Mtb)
  • Inhibits mitochondrial protein synthesis (MPS) in HepG2 cells
  • Effective in mouse models of methicillin-resistant Staphylococcus aureus infection
  • Used for the study of tuberculosis
Catalog No. 50-003-50329 Supplier Medchemexpress LLC Supplier No. HY13939850MG
TBI-223 is an orally active oxazolidinone antibiotic and an antimicrobial that demonstrates activity against Mycobacterium tuberculosis (Mtb). It exhibits an IC50 of 68 μg/mL for inhibiting mitochondrial protein synthesis (MPS) in HepG2 cells. It has shown effectiveness in three mouse models of methicillin-resistant Staphylococcus aureus infection, and can be utilized for the study of tuberculosis.
  • Orally active oxazolidinone antibiotic
  • Antimicrobial activity against Mycobacterium tuberculosis (Mtb)
  • Inhibits mitochondrial protein synthesis (MPS) in HepG2 cells
  • Effective in mouse models of methicillin-resistant Staphylococcus aureus infection
  • Used for the study of tuberculosis