Learn More
Medchemexpress LLC TBI-223 | 2071265-08-0 | 99.7% | 365.36 g/mol | 100 MG
SDP
Catalog No. 5000352380
Encompass_Preferred
TBI-223 is an orally active oxazolidinone antibiotic and an antimicrobial. TBI-223 shows activity against Mycobacterium tuberculosis (Mtb). TBI-223 exhibits an IC50 of 68 μg/mL for inhibiting mitochondrial protein synthesis (MPS) in HepG2 cells. TBI-223 is effective in three mouse models (bloodstream infection, skin infection, and bone infection) of methicillin-resistant staphylococcus aureus infection. TBI-223 can be used for the study of tuberculosis.
  • Orally active oxazolidinone antibiotic
  • Antimicrobial
  • Active against Mycobacterium tuberculosis (Mtb)
  • Exhibits an IC50 of 68 μg/mL for inhibiting mitochondrial protein synthesis (MPS) in HepG2 cells
  • Effective in three mouse models of methicillin-resistant staphylococcus aureus infection
  • Can be used for the study of tuberculosis
Catalog No. 50-003-52380 Supplier Medchemexpress LLC Supplier No. HY139398100MG
TBI-223 is an orally active oxazolidinone antibiotic and an antimicrobial. TBI-223 shows activity against Mycobacterium tuberculosis (Mtb). TBI-223 exhibits an IC50 of 68 μg/mL for inhibiting mitochondrial protein synthesis (MPS) in HepG2 cells. TBI-223 is effective in three mouse models (bloodstream infection, skin infection, and bone infection) of methicillin-resistant staphylococcus aureus infection. TBI-223 can be used for the study of tuberculosis.
  • Orally active oxazolidinone antibiotic
  • Antimicrobial
  • Active against Mycobacterium tuberculosis (Mtb)
  • Exhibits an IC50 of 68 μg/mL for inhibiting mitochondrial protein synthesis (MPS) in HepG2 cells
  • Effective in three mouse models of methicillin-resistant staphylococcus aureus infection
  • Can be used for the study of tuberculosis