Learn More
Sino Biological Recombinant Human STYK1/NOK Protein (GST Tag)
Catalog No. 502572990
Encompass_Preferred
Recombinant human STYK1(NOK) (58-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-2990 Supplier Sino Biological Supplier No. S5335G1000
Recombinant human STYK1(NOK) (58-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.