Learn More
Medchemexpress LLC TBK1/IKKε-IN-4 | 1381930-17-1 | 99.8% | 533.62 g·mol⁻¹ | C28H35N7O4 | 50MG
SDP
Catalog No. 5000236346
Encompass_Preferred
TBK1/IKKε-IN-4 is a 6-aminopyrazolopyrimidine derivative that potently and selectively inhibits TANK-binding kinase 1 (TBK1) and IκB kinase-ε (IKKε). Supplied as a research reagent, it is characterized for potency and purity and is available in multiple milligram-scale pack sizes for laboratory studies.
  • Potent dual TBK1 and IKKε inhibitor with IC50 values of 13 nM (TBK1) and 59 nM (IKKε).
  • High purity: 99.8%.
  • Molecular weight 533.62 g·mol⁻¹; formula C28H35N7O4.
  • Available in multiple pack sizes including 50 mg for small-scale studies.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-36346 Supplier Medchemexpress LLC Supplier No. HY12465250MG
Add to Cart
Add to Cart
TBK1/IKKε-IN-4 is a 6-aminopyrazolopyrimidine derivative that potently and selectively inhibits TANK-binding kinase 1 (TBK1) and IκB kinase-ε (IKKε). Supplied as a research reagent, it is characterized for potency and purity and is available in multiple milligram-scale pack sizes for laboratory studies.
  • Potent dual TBK1 and IKKε inhibitor with IC50 values of 13 nM (TBK1) and 59 nM (IKKε).
  • High purity: 99.8%.
  • Molecular weight 533.62 g·mol⁻¹; formula C28H35N7O4.
  • Available in multiple pack sizes including 50 mg for small-scale studies.
  • Intended for research use only; not for human therapeutic use.