Learn More
Medchemexpress LLC HY-128679 5mg Medchemexpress, TBK1/IKKε-IN-5 CAS:1893397-65-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1286795MG
Encompass_Preferred
Medchemexpress, HY-128679 5mg TBK1/IKKε-IN-5 CAS:1893397-65-3 TBK1/IKKε-IN-5 (compound 1) is a dual TBK1 and IKKε inhibitor, with IC 50 values of 1 nM and 5.6 nM for TBK1 and IKKε, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2628