Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tbk1/ikkepsilon-in-4 | 1381930-17-1 | 99.4% | 533.62 g·mol-1 | C28H35N7O4 | 10MG
SDP

Catalog No. 5000237014
Encompass_Preferred

TBK1/IKKε-IN-4 is a 6-aminopyrazolopyrimidine-derived small-molecule inhibitor selective for TBK1 and IKKε (CAS 1381930-17-1), supplied as a solid research reagent for signaling pathway and preclinical studies.

  • Potent, selective inhibitor of TBK1 and IKKε for pathway interrogation.
  • Chemical formula C28H35N7O4; molecular weight ≈ 533.62 g·mol-1.
  • High purity reported on analysis (≈99.4%).
  • Storage: solid at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month), protected from moisture and light.
  • Intended for research use in signaling and cancer biology studies.

Catalog No. 50-002-37014 Supplier Medchemexpress LLC Supplier No. HY12465210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TBK1/IKKε-IN-4 is a 6-aminopyrazolopyrimidine-derived small-molecule inhibitor selective for TBK1 and IKKε (CAS 1381930-17-1), supplied as a solid research reagent for signaling pathway and preclinical studies.

  • Potent, selective inhibitor of TBK1 and IKKε for pathway interrogation.
  • Chemical formula C28H35N7O4; molecular weight ≈ 533.62 g·mol-1.
  • High purity reported on analysis (≈99.4%).
  • Storage: solid at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month), protected from moisture and light.
  • Intended for research use in signaling and cancer biology studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.