Learn More
Aviva Systems Biology Corp TBL1Y Antibody - middle region (ARP51691_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51691_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBL1Y. It was validated on Western Blot. TBL1Y,transducin (beta)-like 1, Y-linked,TBL1; TBL1Y,F-box-like/WD repeat-containing protein TBL1Y,F-box-like/WD repeat-containing protein TBL1Y,transducin beta-like 1,transducin beta-like protein 1Y,transducin-beta-like protein 1, Y-linkedThe immunogen is a synthetic peptide directed towards the middle region of human TBL1Y. Alternate names: TBL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-584