Learn More
Uline Tabletop Impulse Sealer with Cutter - 16, Aluminum
Catalog No. NC1715554
Encompass
Seal and trim poly tubing in one step, Press gently to seal and slide cutter across tubing to make exact bag size, 8-step adjustable timer, Replacement Blades and Service Kit with replacement heating element available, Cuts 1/4 above the seal, Handles 8 - 10 seals per minute, UL Listed, 670 W, 6 amps
Catalog No. NC1715554 Supplier Uline Supplier No. H963
Seal and trim poly tubing in one step, Press gently to seal and slide cutter across tubing to make exact bag size, 8-step adjustable timer, Replacement Blades and Service Kit with replacement heating element available, Cuts 1/4 above the seal, Handles 8 - 10 seals per minute, UL Listed, 670 W, 6 amps