Learn More
Maine Molecular Quality Controls Inc INTROL TBNEG-30
SDP
Supplier:  Maine Molecular Quality Controls Inc M115
Encompass
INTROL TBNEG-30 is intended for use as a quality control to monitor analytical performance of the M. tuberculosis (MTB) assay on the GeneXpert System. INTROL TBNEG-30 is formulated to monitor the detection of MTB and multi-drug resistant MTB (MDR-TB) mutations. INTROL TBNEG-30 is comprised of 30 bottles, 1 mL each, containing buffer and preservative only, no MTB DNA, no cells.
Catalog No. NC2875400