Learn More
AVANTOR MASTERFLEX (VWR LLC) Catalyst Pump Tubing, Platinum-Cured Silicone, 6.4 mm ID, 2.4 mm wall, 15 m
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9700077
Encompass
Catalyst pump tubing comes in four formulations to cover most chemical compatible applications. FH100X and FH100DX utilize tubing with a wall thickness of 2.4 mm. Standard 15 meter lengths.
Catalog No. NC0978198
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.