Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, Gore STA-PURE Series PFL, 3.2 mm ID x 8 mm OD; 355 mm L
Catalog No. NC1633606
Encompass
Applications: Chemical and pharmaceutical applications. Characteristics: High resilience and strength, low spallation and gas permeability. Opaque. Certifications: USP Class VI compliant, manufactured in Class 7 (ISO 146441-1) cleanroom, under cGMP certified to ISO 13485 and 15378. Temperature range: 0 to 150¿C (Series PFL) Sterilize: Autoclave, CIP/SIP Series PFL tubing is designed to withstand pumping aggressive chemicals with a composite structure made of platinum-cured perfluoroelastomer and ePTFE. Long-term testing showed pumping common organic solvents (acetic acid, acetone, and ethanol) at 10 psig did not reduce the pumping life of the tubing compared with pumping deionized water at the same pressure.
Catalog No. NC1633606 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9625224
Applications: Chemical and pharmaceutical applications. Characteristics: High resilience and strength, low spallation and gas permeability. Opaque. Certifications: USP Class VI compliant, manufactured in Class 7 (ISO 146441-1) cleanroom, under cGMP certified to ISO 13485 and 15378. Temperature range: 0 to 150¿C (Series PFL) Sterilize: Autoclave, CIP/SIP Series PFL tubing is designed to withstand pumping aggressive chemicals with a composite structure made of platinum-cured perfluoroelastomer and ePTFE. Long-term testing showed pumping common organic solvents (acetic acid, acetone, and ethanol) at 10 psig did not reduce the pumping life of the tubing compared with pumping deionized water at the same pressure.