Learn More
AVANTOR MASTERFLEX (VWR LLC) TBNG LMT-55 0.51MM ID 33FT/PK
Catalog No. 502743013
Encompass
Masterflex Ismatec Pump Tubing, Tygon LMT-55, 0.51 mm ID; 33 ft, Avantor
Catalog No. 50-274-3013 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0550218
Masterflex Ismatec Pump Tubing, Tygon LMT-55, 0.51 mm ID; 33 ft, Avantor