Learn More
AVANTOR MASTERFLEX (VWR LLC) C-Flex ULTRA tubing, I/P 73, 25 ft.
Catalog No. NC0925495
Encompass
Masterflex C-Flex ULTRA tubing, I/P 73, 25 ft., Long life tubing for any Masterflex pump used in your pharma and bioprocess applications, Applications: General-purpose lab uses requiring high performance and long life.Characteristics: Exceptional tensile and tear strength; good biocompatibility; low spallation, reduced residue and low gas permeability; and a smooth surface. Translucent.
Catalog No. NC0925495 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0643473
Masterflex C-Flex ULTRA tubing, I/P 73, 25 ft., Long life tubing for any Masterflex pump used in your pharma and bioprocess applications, Applications: General-purpose lab uses requiring high performance and long life.Characteristics: Exceptional tensile and tear strength; good biocompatibility; low spallation, reduced residue and low gas permeability; and a smooth surface. Translucent.