Learn More
Accuflow Systems TBNG MFLEX FOOD A60F#16 500FT
Catalog No. 501109002
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Spooled Masterflex Norprene Food tubing, L/S 16, 500 ft. Similar to Norprene, with FDA compliant materials Excellent for food and dairy applications Good flow consistency Heat and ozone resistant Heat
Catalog No. 50-110-9002 Supplier Accuflow Systems Supplier No. 0641516
Manufactured by Cole Parmer and sold by Accuflow. Spooled Masterflex Norprene Food tubing, L/S 16, 500 ft. Similar to Norprene, with FDA compliant materials Excellent for food and dairy applications Good flow consistency Heat and ozone resistant Heat