Learn More
Accuflow Systems TBNG MFLEX FOOD A60F #16 50FT
Catalog No. 501108845
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex Norprene Food tubing (A60 F), L/S 16, 50 ft. Applications: Ideal for the food, dairy, and beverage industries. Characteristics: Heat, ozone, and UV light resistant; nonaging; nonoxidizing; super
Catalog No. 50-110-8845 Supplier Accuflow Systems Supplier No. 0640216
Manufactured by Cole Parmer and sold by Accuflow. Masterflex Norprene Food tubing (A60 F), L/S 16, 50 ft. Applications: Ideal for the food, dairy, and beverage industries. Characteristics: Heat, ozone, and UV light resistant; nonaging; nonoxidizing; super