Learn More
Accuflow Systems TBNG MFLEX SILICONE #14 500FT
Supplier:  Accuflow Systems 9640314
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Spooled Masterflex platinum-cured silicone tubing, L/S 14, 500 ft. Slightly greater clarity Smooth surface; low protein binding levels Preferred by pharmaceutical & biotechnology industries Fewer potentia
Catalog No. 50-110-8968