Learn More
Accuflow Systems TBNG MFLEX SILICONE #15 400FT
Supplier:  Accuflow Systems 9640315
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Spooled Masterflex platinum-cured silicone tubing, L/S 15, 400 ft. Slightly greater clarity Smooth surface; low protein binding levels Preferred by pharmaceutical & biotechnology industries Fewer potential
Catalog No. 50-110-8978