Learn More
Accuflow Systems TBNG MFLEX SILICONE #73 100FT
Catalog No. 501108980
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Spooled Masterflex BioPharm silicone tubing, I/P 73, 100 ft. Platinum-cured silicone Ultra-smooth inner surface Fewer extractables Minimizes particle entrapment Perfect for lab, biotechnology, & pharmaceut
Catalog No. 50-110-8980 Supplier Accuflow Systems Supplier No. 9642373
Manufactured by Cole Parmer and sold by Accuflow. Spooled Masterflex BioPharm silicone tubing, I/P 73, 100 ft. Platinum-cured silicone Ultra-smooth inner surface Fewer extractables Minimizes particle entrapment Perfect for lab, biotechnology, & pharmaceut