Learn More
Accuflow Systems TBNG MFLEX TYGON E-LAB#16 50FT
Catalog No. 501108859
Encompass_Preferred
Manufactured by Cole Parmer & sold by Accuflow. Masterflex Tygon E-Lab (E3603) Pump Tubing, L/S 16, 50 ft. Specifications: Tubing formulation - Tygon E-Lab (E-3603) Pump Series - L/S Masterflex tubing size (L/S) - 16 Tubing ID (mm) - 3.1 Hose barb size (i
Catalog No. 50-110-8859 Supplier Accuflow Systems Supplier No. 0650916
Manufactured by Cole Parmer & sold by Accuflow. Masterflex Tygon E-Lab (E3603) Pump Tubing, L/S 16, 50 ft. Specifications: Tubing formulation - Tygon E-Lab (E-3603) Pump Series - L/S Masterflex tubing size (L/S) - 16 Tubing ID (mm) - 3.1 Hose barb size (i