Learn More
Accuflow Systems TBNG MFLEX TYGON E-LAB#17 50FT
Catalog No. 501108838
Encompass_Preferred
Manufactured by Cole Parmer & sold by Accuflow. Masterflex Tygon E-Lab (E-3603) Pump Tubing, L/S 17, 50 ft. Specifications: Product Type Pump Tubing Tubing formulation - Tygon E-Lab (E-3603) Pump Series - L/S Masterflex tubing size (L/S) - 17 Tubing ID (
Catalog No. 50-110-8838 Supplier Accuflow Systems Supplier No. 0650917
Manufactured by Cole Parmer & sold by Accuflow. Masterflex Tygon E-Lab (E-3603) Pump Tubing, L/S 17, 50 ft. Specifications: Product Type Pump Tubing Tubing formulation - Tygon E-Lab (E-3603) Pump Series - L/S Masterflex tubing size (L/S) - 17 Tubing ID (