Learn More
AVANTOR MASTERFLEX (VWR LLC) TBNG MFLEX TYGON ELFL 35 25FT
Catalog No. 502742909
Encompass
Masterflex L/S High-Performance Precision Pump Tubing, Tygon S3™ E-LFL , L/S 35; 25 ft, Avantor
Catalog No. 50-274-2909 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0644035
Masterflex L/S High-Performance Precision Pump Tubing, Tygon S3™ E-LFL , L/S 35; 25 ft, Avantor