Learn More
AVANTOR MASTERFLEX (VWR LLC) TBNG MFLEX TYGON ELFL 82 25FT
Catalog No. 502742905
Encompass
Masterflex I/P Precision Pump Tubing, Tygon S3™ E-LFL, I/P 82; 25 ft, Avantor
Catalog No. 50-274-2905 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0644082
Masterflex I/P Precision Pump Tubing, Tygon S3™ E-LFL, I/P 82; 25 ft, Avantor