Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P High-Performance Precision Pump Tubing, Tygon E-LFL, I/P 88; 25 ft
Catalog No. NC1614525
Encompass
Masterflex Tygon E-FLF High-Performance Precision pump tubing is a non-DEHP formulation specially formulated for even longer pumping life with superior flexibility. This tubing features low particulate spallation and no phthalates or BPA, protecting fluid purity. Ideal for food and beverage, cosmetics, and pharmaceutical applications, this tubing complies with FDA, USP Class VI, and EU Foods standards.
Catalog No. NC1614525 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0644088/DEL
Masterflex Tygon E-FLF High-Performance Precision pump tubing is a non-DEHP formulation specially formulated for even longer pumping life with superior flexibility. This tubing features low particulate spallation and no phthalates or BPA, protecting fluid purity. Ideal for food and beverage, cosmetics, and pharmaceutical applications, this tubing complies with FDA, USP Class VI, and EU Foods standards.