Learn More
Accuflow Systems TBNG MFLEX TYGON F&L #13 50FT
Catalog No. 501109000
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex Tygon Fuel & Lubricant tubing, L/S 13, 50 ft. Ideal for transferring hydrocarbons, gasoline, kerosene, heating oils, cutting compounds, and glycol based coolants Applications: Fuels and industr
Catalog No. 50-110-9000 Supplier Accuflow Systems Supplier No. 0640113
Manufactured by Cole Parmer and sold by Accuflow. Masterflex Tygon Fuel & Lubricant tubing, L/S 13, 50 ft. Ideal for transferring hydrocarbons, gasoline, kerosene, heating oils, cutting compounds, and glycol based coolants Applications: Fuels and industr