Learn More
AVANTOR MASTERFLEX (VWR LLC) TBNG TYGON 2001 2.06MM ID 33FT
Catalog No. 502741884
Encompass
Masterflex Ismatec Pump Tubing, Versilon™ 2001, 2.06 mm ID x 3.76 mm OD; 33 ft, Avantor
Catalog No. 50-274-1884 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9646742
Masterflex Ismatec Pump Tubing, Versilon™ 2001, 2.06 mm ID x 3.76 mm OD; 33 ft, Avantor