Learn More
Elemental Scientific Inc TBNG TYGON.51MM YEL/OR 12PK
SDP
Catalog No. NC9752720
Encompass_Preferred
Peristaltic Pump Tubing, PVC, 2-stop, 0.51mm ID, 2.0mm flared 151mm bridge distance (orange/yellow), 12 pc.
This item is not returnable. View return policy
Catalog No. NC9752720 Supplier Elemental Scientific Inc Supplier No. ES43971051
Peristaltic Pump Tubing, PVC, 2-stop, 0.51mm ID, 2.0mm flared 151mm bridge distance (orange/yellow), 12 pc.