Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Precision Pump Tubing, FDA-Compliant Viton, I/P 73; 25 ft
Catalog No. NC9633974
Encompass
Masterflex FDA-Compliant Viton Precision pump tubing is designed for enhanced chemical compatibility at elevated temperatures. Tubing tolerates the transfer of the harshest chemicals such as corrosive solvents and aliphatic and aromatic hydrocarbons. The ultra-low gas permeability maintains integrity of fluid while protecting the user and environment. Ideal for where FDA compliance and superior chemical compatibility are required.
Catalog No. NC9633974 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9641273
Masterflex FDA-Compliant Viton Precision pump tubing is designed for enhanced chemical compatibility at elevated temperatures. Tubing tolerates the transfer of the harshest chemicals such as corrosive solvents and aliphatic and aromatic hydrocarbons. The ultra-low gas permeability maintains integrity of fluid while protecting the user and environment. Ideal for where FDA compliance and superior chemical compatibility are required.