Learn More
ACCELLIX INC TBNK-16 RTF ASAY
Catalog No. NC2542290
Encompass
  • Automated workflow streamlines the entire GMP cell phenotyping process from sample preparation to data acquisition and analysis.
  • Ready-to-use cartridge minimizes manual intervention and potential for human error.
  • Reagents are stable at ambient temperatures, simplifying storage requirements.
  • Integrated quality control includes control beads for accurate cell counting.
  • Comprehensive lymphocyte profiling provides detailed immune cell analysis.
  • Lysis buffer included, suitable for apheresis and whole blood samples.
  • Sample compatibility: Apheresis, whole blood, RBC-containing samples.
  • Sample concentration range: 1-50 million cells/mL.
  • Automation for sample preparation, acquisition, and analysis.
  • Simple workflow, streamlined via QR code.
  • In-run quality control ensures robust assay performance.
  • Result output includes frequencies and counts.
Catalog No. NC2542290 Supplier ACCELLIX INC Supplier No. A10031L
  • Automated workflow streamlines the entire GMP cell phenotyping process from sample preparation to data acquisition and analysis.
  • Ready-to-use cartridge minimizes manual intervention and potential for human error.
  • Reagents are stable at ambient temperatures, simplifying storage requirements.
  • Integrated quality control includes control beads for accurate cell counting.
  • Comprehensive lymphocyte profiling provides detailed immune cell analysis.
  • Lysis buffer included, suitable for apheresis and whole blood samples.
  • Sample compatibility: Apheresis, whole blood, RBC-containing samples.
  • Sample concentration range: 1-50 million cells/mL.
  • Automation for sample preparation, acquisition, and analysis.
  • Simple workflow, streamlined via QR code.
  • In-run quality control ensures robust assay performance.
  • Result output includes frequencies and counts.