Learn More
Medchemexpress LLC TBOPP | 1996629-79-8 | 99.6% | 490.49 | C24H21F3N2O4S | 50MG
SDP
Catalog No. 5000236329
Encompass_Preferred
TBOPP is a small-molecule research reagent that selectively inhibits DOCK1. It blocks DOCK1-mediated Rac activation with an IC50 of 8.4 μM and binds the DOCK1 DHR-2 domain with a Kd of 7.1 μM. TBOPP has shown anti-tumor activity in preclinical studies and is provided as a high-purity compound with available powder and DMSO solution formats for biochemical and cellular experiments.
  • High purity suitable for research applications.
  • Selective inhibitor of DOCK1 with IC50 8.4 μM and Kd 7.1 μM.
  • Demonstrated anti-tumor activity in preclinical models.
  • Available as powder and as a DMSO solution for flexible dosing.
  • Comes with supporting documentation such as datasheet, COA, and SDS.
  • Stable under recommended cold storage for long-term preservation.
Catalog No. 50-002-36329 Supplier Medchemexpress LLC Supplier No. HY12471150MG
TBOPP is a small-molecule research reagent that selectively inhibits DOCK1. It blocks DOCK1-mediated Rac activation with an IC50 of 8.4 μM and binds the DOCK1 DHR-2 domain with a Kd of 7.1 μM. TBOPP has shown anti-tumor activity in preclinical studies and is provided as a high-purity compound with available powder and DMSO solution formats for biochemical and cellular experiments.
  • High purity suitable for research applications.
  • Selective inhibitor of DOCK1 with IC50 8.4 μM and Kd 7.1 μM.
  • Demonstrated anti-tumor activity in preclinical models.
  • Available as powder and as a DMSO solution for flexible dosing.
  • Comes with supporting documentation such as datasheet, COA, and SDS.
  • Stable under recommended cold storage for long-term preservation.