Learn More
Sigma Aldrich Fine Chemicals Biosciences TBP TATA box binding prote
Catalog No. 502904042
Encompass
TBP (TATA-box binding protein) was originally recognized as a part of the class II initiation factor TfIID. Its C-terminal region is phylogenetically conserved and is composed of 180-amino acids containing a highly basic segment the basic repeat flanked by two direct repeats. The N-terminal domain of this protein is not conserved across species and differs in number of residues. TBP gene is localized to human chromosome 6.
Catalog No. 50-290-4042 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRP200310UG
TBP (TATA-box binding protein) was originally recognized as a part of the class II initiation factor TfIID. Its C-terminal region is phylogenetically conserved and is composed of 180-amino acids containing a highly basic segment the basic repeat flanked by two direct repeats. The N-terminal domain of this protein is not conserved across species and differs in number of residues. TBP gene is localized to human chromosome 6.