Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich TBPe

Catalog No. 900937250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-093-7250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-093-7250MG Supplier Sigma Aldrich Supplier No. 900937250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Electron transport and hole blocking material in organic light emitting diodes (OLED).

Specifications

CAS 80663-92-9
Molecular Formula C36H44
Linear Formula C36H44
Quantity 250 mg
MDL Number MFCD09835379
Synonym 2,5,8,11-Tetra-tert-butylperylene; 2,5,8,11-Tetrakis(1,1-dimethylethyl)perylene; TBP
Molecular Weight (g/mol) 476.73
Percent Purity 99% (sublimed, HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.