Learn More
Aviva Systems Biology Corp TBPL1 Antibody - middle region (ARP35751_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35751_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TBPL1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human TBPL1. Alternate names: TLF, TLP, STUD, TRF2, MGC:8389, MGC:9620ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53019