Learn More
World Precision Instrument TBR NO Sensor Calibration Solution
SDP
Catalog No. 502538489
Encompass
  • Using CuCl2 as a catalyst is convenient method to calibrate your NO micro-sensor.
  • This solution is the one used in WPI laboratories.
  • It provides reliable data of calibration.
  • Volume of 125 mL
  • Package of 5
Catalog No. 50-253-8489 Supplier World Precision Instrument Supplier No. TBRNOCAL
  • Using CuCl2 as a catalyst is convenient method to calibrate your NO micro-sensor.
  • This solution is the one used in WPI laboratories.
  • It provides reliable data of calibration.
  • Volume of 125 mL
  • Package of 5