Learn More
World Precision Instrument TBR PBS Calibration Solution
SDP
Catalog No. 502538488
Encompass
  • Before attempting to use your sensor in an experimental solution, calibrate it under standard conditions to verify proper operation and performance.
  • This solution is the one used in WPI laboratories.
  • Volume of 125 mL
  • Package of 5
Catalog No. 50-253-8488 Supplier World Precision Instrument Supplier No. TBRPBSCAL
  • Before attempting to use your sensor in an experimental solution, calibrate it under standard conditions to verify proper operation and performance.
  • This solution is the one used in WPI laboratories.
  • Volume of 125 mL
  • Package of 5