Learn More
Aviva Systems Biology Corp TBR1 Antibody - middle region (ARP31497_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31497_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TBR1. Alternate names: MGC141978, TES-56, TBR-1ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig
Catalog No. 50-014-54245