Learn More
Aviva Systems Biology Corp Tbr1 Antibody - N-terminal region (ARP36982_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36982_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tbr1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of mouse Tbr1. Alternate names: -ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-558