Learn More
Aviva Systems Biology Corp TBRG1 Antibody - N-terminal region (ARP60647_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60647_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBRG1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TBRG1. Alternate names: FLJ14621, FLJ25020, FLJ90113, MGC129890, NIAM, TB-5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-114