Learn More
ABclonal Technology TBRG4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A157531000UL
Encompass_Preferred
Enables RNA binding activity. Involved in mitochondrial mRNA processing and regulation of mitochondrial mRNA stability. Located in mitochondrial matrix.
Catalog No. 50-001-06480