Learn More
ABclonal Technology TBRG4 Rabbit pAb
SDP
Catalog No. 5000108012
Encompass_Preferred
Enables RNA binding activity. Involved in mitochondrial mRNA processing and regulation of mitochondrial mRNA stability. Located in mitochondrial matrix.
Catalog No. 50-001-08012 Supplier ABclonal Technology Supplier No. A15753500UL
Enables RNA binding activity. Involved in mitochondrial mRNA processing and regulation of mitochondrial mRNA stability. Located in mitochondrial matrix.