Learn More
Biotang Inc TRIS-Buffered Saline (10X, High Salt) Tris 0.5M NaCl 4.5M , 1000 ml
Catalog No. 50155868 Shop All BIOTANG Inc Products
Encompass_Preferred
TRIS-Buffered Saline (10X, High Salt) Tris 0.5M NaCl 4.5M , 1000 ml.
Catalog No. 50-155-868 Supplier Biotang Inc Supplier No. BTBB546
TRIS-Buffered Saline (10X, High Salt) Tris 0.5M NaCl 4.5M , 1000 ml.