Learn More
Biotang Inc TRIS-Buffered Saline (TBS, 10X) For Western Blot Washing; (NaCl 1.5M Tris 0.5M), 1000 ml
Catalog No. 50155866 Shop All BIOTANG Inc Products
Encompass_Preferred
TRIS-Buffered Saline (TBS, 10X) For Western Blot Washing; (NaCl 1.5M Tris 0.5M), 1000 ml.
Catalog No. 50-155-866 Supplier Biotang Inc Supplier No. BTBB544
TRIS-Buffered Saline (TBS, 10X) For Western Blot Washing; (NaCl 1.5M Tris 0.5M), 1000 ml.