Learn More
AG Scientific Inc TBS 10X Solution 1 L
Catalog No. 502268422
Encompass_Preferred
TRIS Buffered Saline is a buffer used in some biochemical techniques for pH maintenance within a narrow range.pH 7.25-7.55
Catalog No. 50-226-8422 Supplier AG Scientific Inc Supplier No. T27471L
TRIS Buffered Saline is a buffer used in some biochemical techniques for pH maintenance within a narrow range.pH 7.25-7.55