Learn More
Boston Bioproducts Inc Tris Buffered Saline, TBS (20X, for Western Blot Washing) - 4L
Catalog No. NC0287249
Encompass
The Tris Buffered Saline (TBS) is an isotonic non-toxic buffer solution used commonly in biochemical applications. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. This TBS Solution is formulated at a 20X concentration, and is useful for washing in Western Blotting.
Catalog No. NC0287249 Supplier Boston Bioproducts Inc Supplier No. BM301X4L
The Tris Buffered Saline (TBS) is an isotonic non-toxic buffer solution used commonly in biochemical applications. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. Tris maintains a stable pH range typically between 7.4 to 8.0 while NaCl regulates osmotic balance via its elected concentration. This TBS Solution is formulated at a 20X concentration, and is useful for washing in Western Blotting.