Learn More
Cayman Chemical TBS asy Buffer ConcentrAT 10mL
SDP
Catalog No. 502942201
Encompass_Preferred
An internal standard for the quantification of AEA by GC- or LC-MS.
Catalog No. 50-294-2201 Supplier Cayman Chemical Supplier No. 40008410 ML
An internal standard for the quantification of AEA by GC- or LC-MS.